accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-51-3

IUPAC Name: disodium 3-hydroxynaphthalene-2,7-disulfonate
Synonyms: disodium 3-hydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 348.25
Molecular Formula: C10H6Na2O7S2
InChi Key: VNEBWJSWMVTSHK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=C(C=C2C=C(C=CC2=C1)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results