accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-61-5

IUPAC Name: 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide
Synonyms: 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide
Molecular Weight (g/mol): 277.32
Molecular Formula: C18H15NO2
InChi Key: FBLAHUMENIHUGG-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(=O)C1=C(O)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results