accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-62-6

IUPAC Name: sodium 3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-olate
Synonyms: sodium 3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-olate
Molecular Weight (g/mol): 315.3
Molecular Formula: C18H14NNaO3
InChi Key: OXXQDUUQLWQUEX-UHFFFAOYSA-M
SMILES: [Na+].COC1=CC=CC=C1NC(=O)C1=C([O-])C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results