CAS RN 135-64-8
IUPAC Name:
3-hydroxy-N-(naphthalen-2-yl)naphthalene-2-carboxamide
Synonyms:
3-hydroxy-N-(naphthalen-2-yl)naphthalene-2-carboxamide
Molecular Weight (g/mol):
313.36
Molecular Formula:
C21H15NO2
InChi Key:
PMYDPQQPEAYXKD-UHFFFAOYSA-N
SMILES:
OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=C2C=CC=CC2=C1