accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-68-2

IUPAC Name: 4'-chloro-[1,1'-biphenyl]-4-amine
Synonyms: 4-amino-4'-chlorodiphenyl
Molecular Weight (g/mol): 203.67
Molecular Formula: C12H10ClN
InChi Key: OREQWMWYRYXCDF-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results