accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-70-6

CAS RN 135-70-6

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-1,1'-biphenyl
Synonyms: p-quaterphenyl p-tetraphenyl quadriphenyl
Molecular Weight (g/mol): 306.41
Molecular Formula: C24H18
InChi Key: GPRIERYVMZVKTC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results