accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-77-3

IUPAC Name: 1,2,4-trimethoxybenzene
Synonyms: 1,2,4-trimethoxybenzene
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: AGIQIOSHSMJYJP-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(OC)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)