accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-88-6

IUPAC Name: N-phenylnaphthalen-2-amine
Synonyms: anilinonaphthalene AK 1 (stabilizer) naftam 2 neozone antioxidant 116 nocrac D antioxidant D nonox DN antioxidant pbn PBNA
Molecular Weight (g/mol): 219.29
Molecular Formula: C16H13N
InChi Key: KEQFTVQCIQJIQW-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)