accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiH3CClOH3C

CAS RN 13501-76-3

IUPAC Name: (3-chloropropyl)diethoxymethylsilane
Synonyms: (3-chloropropyl)diethoxymethylsilane
Molecular Weight (g/mol): 210.77
Molecular Formula: C8H19ClO2Si
InChi Key: KEZMLECYELSZDC-UHFFFAOYSA-N
SMILES: CCO[Si](C)(CCCCl)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results