CAS RN 135025-34-2
CAS RN 135025-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4-Cyclohexanedimethanol Dilaurate 90.0+%, TCI America™
CAS: 135025-34-2 Molecular Formula: C32H60O4 MDL Number: MFCD00060060