accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOOOOOOOHNNHNHNNN(R)(S)(R)(R)(S)(R)CH2(S)CH3CH3H3CCH3HHHH

CAS RN 1350514-68-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results