accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13509-27-8

IUPAC Name: methyl phenyl carbonate
Synonyms: methyl phenyl carbonate
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: XTBFPVLHGVYOQH-UHFFFAOYSA-N
SMILES: COC(=O)OC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results