accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OH2OH2OOSOOOOSOOOOSOOOPrPr

CAS RN 13510-41-3

IUPAC Name: dipraseodymium(3+) octahydrate trisulfate
Synonyms: dipraseodymium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 714.1
Molecular Formula: H16O20Pr2S3
InChi Key: NIJXLQIGVGZNKP-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Pr+3].[Pr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)