accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ClOHO

CAS RN 13511-38-1

IUPAC Name: 3-chloro-2,2-dimethylpropanoic acid
Synonyms: 3-chloro-2,2-dimethylpropanoic acid
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: YBJGQSNSAWZZHL-UHFFFAOYSA-N
SMILES: CC(C)(CCl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results