accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13515-76-9

IUPAC Name: methyl 3-hydroxy-2-[(triphenylmethyl)amino]propanoate
Synonyms: methyl 3-hydroxy-2-[(triphenylmethyl)amino]propanoate
Molecular Weight (g/mol): 361.44
Molecular Formula: C23H23NO3
InChi Key: LXAWQKKSNNYYEK-UHFFFAOYNA-N
SMILES: COC(=O)C(CO)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results