accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CHNOOCH3

CAS RN 13515-93-0

IUPAC Name: hydrogen methyl 2-(methylamino)acetate chloride
Synonyms: hydrogen methyl 2-(methylamino)acetate chloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: HQZMRJBVCVYVQA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)