accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2ONaI

CAS RN 13517-06-1

IUPAC Name: sodium dihydrate iodide
Synonyms: sodium dihydrate iodide
Molecular Weight (g/mol): 185.92
Molecular Formula: H4INaO2
InChi Key: VEEIYHUOLCUPSA-UHFFFAOYSA-M
SMILES: O.O.[Na+].[I-]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)