accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFCH3F

CAS RN 1351985-05-1

IUPAC Name: 2-(2,4-difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2,4-difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 254.08
Molecular Formula: C13H17BF2O2
InChi Key: HDAKNAMXPKYUCU-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(B2OC(C)(C)C(C)(C)O2)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results