CAS RN 13520-72-4
IUPAC Name:
fluorobismuthanone
Synonyms:
fluorobismuthanone
Molecular Weight (g/mol):
245.99
Molecular Formula:
BiFH2O
InChi Key:
KHMIIGVSZMFUNW-UHFFFAOYSA-N
SMILES:
F.O=[BiH]
1
–
1
of
1
results
1
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™
CAS: 13520-72-4 Molecular Formula: BiFO Molecular Weight (g/mol): 243.98