accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH3COHH3COOH3COHH3CCa

CAS RN 135236-72-5

IUPAC Name: calcium bis(3-hydroxy-3-methylbutanoate)
Synonyms: calcium bis(β-hydroxyisovalerate)
Molecular Weight (g/mol): 274.33
Molecular Formula: C10H18CaO6
InChi Key: WLJUMPWVUPNXMF-UHFFFAOYSA-L
SMILES: [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)