accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13529-17-4

IUPAC Name: 5-formylfuran-2-carboxylate
Synonyms: 5-formylfuran-2-carboxylate
Molecular Weight (g/mol): 139.09
Molecular Formula: C6H3O4
InChi Key: SHNRXUWGUKDPMA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)