accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135296-80-9

IUPAC Name: trichloroosmium trihydrate
Synonyms: trichloroosmium trihydrate
Molecular Weight (g/mol): 350.63
Molecular Formula: Cl3H6O3Os
InChi Key: UACQLNPCDXDCID-UHFFFAOYSA-K
SMILES: O.O.O.Cl[Os](Cl)Cl
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)