accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClZnH3CNOHNN

CAS RN 13532-96-2

IUPAC Name: zinc(2+) 4-[ethyl(2-hydroxyethyl)amino]benzene-1-diazonium trichloride
Synonyms: zinc(2+) 4-[ethyl(2-hydroxyethyl)amino]benzenediazonium trichloride
Molecular Weight (g/mol): 363.97
Molecular Formula: C10H14Cl3N3OZn
InChi Key: FOZURDCUSZRTCY-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Zn++].CCN(CCO)C1=CC=C(C=C1)[N+]#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results