accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNBr

CAS RN 13534-89-9

IUPAC Name: 2,3-dibromopyridine
Synonyms: 2,3-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: SLMHHOVQRSSRCV-UHFFFAOYSA-N
SMILES: BrC1=CC=CN=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)