accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHBr

CAS RN 13534-99-1

IUPAC Name: 2-amino-3-bromopyridin-1-ium
Synonyms: 2-amino-3-bromopyridin-1-ium
Molecular Weight (g/mol): 174.02
Molecular Formula: C5H6BrN2
InChi Key: RBCARPJOEUEZLS-UHFFFAOYSA-O
SMILES: NC1=[NH+]C=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)