CAS RN 13536-53-3
CAS RN 13536-53-3
IUPAC Name:
praseodymium(3+) tribromide
Synonyms:
praseodymium(3+) tribromide
Molecular Weight (g/mol):
380.62
Molecular Formula:
Br3Pr
InChi Key:
PLKCYEBERAEWDR-UHFFFAOYSA-K
SMILES:
[Br-].[Br-].[Br-].[Pr+3]
1
–
1
of
1
results
1
Thermo Scientific Chemicals Praseodymium(III) bromide, ultra dry, 99.99% (REO)
CAS: 13536-53-3 MDL Number: MFCD00049762