accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13536-79-3

IUPAC Name: lanthanum(3+) tribromide
Synonyms: lanthanum(III) bromide
Molecular Weight (g/mol): 378.62
Molecular Formula: Br3La
InChi Key: XKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[La+3]
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)