accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClTm

CAS RN 13537-18-3

IUPAC Name: thulium(3+) trichloride
Synonyms: thulium(III) chloride
Molecular Weight (g/mol): 275.28
Molecular Formula: Cl3Tm
InChi Key: ILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Tm+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results