CAS RN 13545-04-5
IUPAC Name:
(2R,3S)-2,3-dimethylbutanedioate
Synonyms:
(2R,3S)-2,3-dimethylbutanedioate
Molecular Weight (g/mol):
144.13
Molecular Formula:
C6H8O4
InChi Key:
KLZYRCVPDWTZLH-ZXZARUISSA-L
SMILES:
C[C@@H]([C@@H](C)C([O-])=O)C([O-])=O
1
–
1
of
1
results
1
Sigma Aldrich 2,3-Dimethylsuccinic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.