accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3ONH(S)(R)HO(R)HONHO(S)(S)HONO(S)NHO(S)NHO(S)(R)OHNO(S)NHO(S)H3COH(S)OH(S)HOOH(S)HONH2O

CAS RN 135575-42-7

IUPAC Name: N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S)-3-[(1S)-2-carbamoyl-1-hydroxyethyl]-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,21,25-tetrahydroxy-15-[(1S)-1-hydroxyethyl]-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexaazatricyclo[22.3.0.0⁹,¹³]heptacosan-18-yl]-10,12-dimethyltetradecanamide
Synonyms: pneumocandin bo
Molecular Weight (g/mol): 1065.23
Molecular Formula: C50H80N8O17
InChi Key: DQXPFAADCTZLNL-YSHLLHMASA-N
SMILES: CCC(C)CC(C)CCCCCCCCC(=O)N[C@H]1C[C@@H](O)[C@@H](O)NC(=O)[C@@H]2[C@@H](O)CCN2C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]2C[C@@H](O)CN2C(=O)[C@@H](NC1=O)[C@H](C)O)[C@H](O)[C@@H](O)C1=CC=C(O)C=C1)[C@@H](O)CC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)