accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2OH2NOHO(?)

CAS RN 13558-31-1

IUPAC Name: 3,6-diamino-9-(2-carboxyphenyl)-10λ⁴-xanthen-10-ylium chloride
Synonyms: rhodamine 110
Molecular Weight (g/mol): 366.8
Molecular Formula: C20H15ClN2O3
InChi Key: MYIOYATURDILJN-UHFFFAOYSA-N
SMILES: [Cl-].NC1=CC2=[O+]C3=CC(N)=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)