CAS RN 135592-13-1
CAS RN 135592-13-1
IUPAC Name:
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Synonyms:
(2R)-2-[(tert-butoxycarbonyl)amino]-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Molecular Weight (g/mol):
491.65
Molecular Formula:
C29H33NO4S
InChi Key:
GFULWLRDJMLHCD-XMMPIXPASA-N
SMILES:
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech Boc-Pen Trt -OH, Advanced ChemTech
CAS: 135592-13-1 Molecular Formula: C29H33NO4S Molecular Weight (g/mol): 491.65 MDL Number: MFCD00237381 InChI Key: GFULWLRDJMLHCD-XMMPIXPASA-N IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid SMILES: CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1