accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1356113-92-2

IUPAC Name: 1,3-diiodo-2-methoxy-5-(trifluoromethyl)benzene
Synonyms: 1,3-diiodo-2-methoxy-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 427.93
Molecular Formula: C8H5F3I2O
InChi Key: XBMZQHBOGXDLDX-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(C=C1I)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results