CAS RN 135613-33-1
IUPAC Name:
ethyl 3-(2-bromophenyl)propanoate
Synonyms:
ethyl 3-(2-bromophenyl)propanoate
Molecular Weight (g/mol):
257.13
Molecular Formula:
C11H13BrO2
InChi Key:
ZKICRDRBPPOQLM-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCC1=CC=CC=C1Br
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-(2-bromophenyl)propionate, 98%, Thermo Scientific Chemicals
Ethyl 3-(2-bromophenyl)propionate, 98%, Thermo Scientific Chemicals
CAS: 135613-33-1 Molecular Formula: C11H13BrO2 Molecular Weight (g/mol): 257.127 MDL Number: MFCD07772962 InChI Key: ZKICRDRBPPOQLM-UHFFFAOYSA-N Synonym: ethyl 3-2-bromophenyl propanoate,3-2-bromo-phenyl-propionic acid ethyl ester,3-2-bromophenyl propionic acid ethyl ester,benzenepropanoic acid, 2-bromo-, ethyl ester,ksc497c5h,ethyl3-2-bromophenyl propanoate,ethyl 2-bromo-3-benzenepropanoate,2-bromo-hydrocinnamic acid ethyl ester,3-2-bromophenyl propanoic acid ethyl ester PubChem CID: 10611137 IUPAC Name: ethyl 3-(2-bromophenyl)propanoate SMILES: CCOC(=O)CCC1=CC=CC=C1Br