accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COOOCH3

CAS RN 13562-81-7

IUPAC Name: pentan-3-yl 3-oxobutanoate
Synonyms: pentan-3-yl 3-oxobutanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: PFTIWTQFHWICDR-UHFFFAOYSA-N
SMILES: CCC(CC)OC(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results