accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3O

CAS RN 13562-84-0

IUPAC Name: hexyl 3-oxobutanoate
Synonyms: hexyl 3-oxobutanoate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: QNZLAXONNWOLJY-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results