accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13565-38-3

IUPAC Name: (2E)-3-(4-fluorophenyl)-1-(4-methylphenyl)prop-2-en-1-one
Synonyms: (2E)-3-(4-fluorophenyl)-1-(4-methylphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 240.28
Molecular Formula: C16H13FO
InChi Key: LUUPODNGYDYQLY-IZZDOVSWSA-N
SMILES: CC1=CC=C(C=C1)C(=O)\C=C\C1=CC=C(F)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results