accessibility menu, dialog, popup

UserName

Viewing profile as user:
PdOSOOO

CAS RN 13566-03-5

IUPAC Name: palladium(2+) sulfate
Synonyms: palladium sulfate
Molecular Weight (g/mol): 202.48
Molecular Formula: O4PdS
InChi Key: RFLFDJSIZCCYIP-UHFFFAOYSA-L
SMILES: [Pd++].[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results