accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13568-40-6

IUPAC Name: dilithium(1+) dioxomolybdenumbis(olate)
Synonyms: lithium molybdate
Molecular Weight (g/mol): 173.83
Molecular Formula: Li2MoO4
InChi Key: NMHMDUCCVHOJQI-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O-][Mo]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)