accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClON(R)(S)NH

CAS RN 135729-62-3

IUPAC Name: hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),9,11-trien-2-one chloride
Synonyms: hydrogen palonosetron chloride
Molecular Weight (g/mol): 332.87
Molecular Formula: C19H25ClN2O
InChi Key: OLDRWYVIKMSFFB-KPVRICSOSA-N
SMILES: [H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)