accessibility menu, dialog, popup

UserName

Viewing profile as user:
GeH4HIIH

CAS RN 13573-08-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 332.49
Molecular Formula: GeH6I2
InChi Key: PUZROLYCAWSHNL-UHFFFAOYSA-N
SMILES: [GeH4].I.I
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)