accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OONHNNHOONCH3CH3OONH3CCH3

CAS RN 135734-39-3

IUPAC Name: N-heptyl-N'-(8-{2-[heptyl(methyl)carbamoyl]-N-(8-{2-[heptyl(methyl)carbamoyl]acetamido}octyl)acetamido}octyl)-N-methylpropanediamide
Synonyms: N-heptyl-N'-(8-{2-[heptyl(methyl)carbamoyl]-N-(8-{2-[heptyl(methyl)carbamoyl]acetamido}octyl)acetamido}octyl)-N-methylpropanediamide
Molecular Weight (g/mol): 863.33
Molecular Formula: C49H94N6O6
InChi Key: JYBGTBZZVHYIJI-UHFFFAOYSA-N
SMILES: CCCCCCCN(C)C(=O)CC(=O)NCCCCCCCCN(CCCCCCCCNC(=O)CC(=O)N(C)CCCCCCC)C(=O)CC(=O)N(C)CCCCCCC
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results