accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHO

CAS RN 135829-04-8

IUPAC Name: benzyl 2-(hydroxymethyl)-2,3-dihydro-1H-indole-1-carboxylate
Synonyms: benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol): 283.33
Molecular Formula: C17H17NO3
InChi Key: QSMOQTIGILELKY-UHFFFAOYNA-N
SMILES: OCC1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results