accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13587-35-4

IUPAC Name: copper(2+) tungsten tetraoxidandiide
Synonyms: copper(2+) tungsten tetraoxidandiide
Molecular Weight (g/mol): 311.39
Molecular Formula: CuO4W
InChi Key: FUZOAUOBBKGKBL-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Cu++].[W]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results