CAS RN 13590-82-4
Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J Synonym: ceric sulfate,cerium iv sulfate,ceric disulfate,ceric sulphate,cerium disulfate,cerium 4+ sulfate,cerium 4+ disulphate,unii-4f66fi5t7x,sulfuric acid, cerium 4+ salt 2:1,cerium 4+ disulfate PubChem CID: 159684 IUPAC Name: λ4-cerium(4+) disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Cerium(IV) sulfate, 95%
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J Synonym: ceric sulfate,cerium iv sulfate,ceric disulfate,ceric sulphate,cerium disulfate,cerium 4+ sulfate,cerium 4+ disulphate,unii-4f66fi5t7x,sulfuric acid, cerium 4+ salt 2:1,cerium 4+ disulfate PubChem CID: 159684 IUPAC Name: cerium(4+);disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Cerium(IV) sulfate, anhydrous, 97%
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J Synonym: ceric sulfate,cerium iv sulfate,ceric disulfate,ceric sulphate,cerium disulfate,cerium 4+ sulfate,cerium 4+ disulphate,unii-4f66fi5t7x,sulfuric acid, cerium 4+ salt 2:1,cerium 4+ disulfate PubChem CID: 159684 IUPAC Name: cerium(4+);disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ceric Sulfate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J PubChem CID: 159684 IUPAC Name: λ⁴-cerium(4+) disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Cerium(IV) sulfate, 0.1N Standardized Solution
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J PubChem CID: 159684 IUPAC Name: cerium(4+);disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ceric Sulfate, Reagent, 85%, Spectrum™ Chemical
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J IUPAC Name: λ⁴-cerium(4+) disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Cerium(IV) sulfate solution, 0.05M Ce(SO4)2, Solstice
CAS: 13590-82-4 Molecular Formula: CeO8S2 Molecular Weight (g/mol): 332.23 MDL Number: MFCD00148852 InChI Key: VZDYWEUILIUIDF-UHFFFAOYSA-J Synonym: ceric sulfate,cerium iv sulfate,ceric disulfate,ceric sulphate,cerium disulfate,cerium 4+ sulfate,cerium 4+ disulphate,unii-4f66fi5t7x,sulfuric acid, cerium 4+ salt 2:1,cerium 4+ disulfate PubChem CID: 159684 IUPAC Name: cerium(4+);disulfate SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Sigma Aldrich Cerium(Iv) Sulfate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.