accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSCH3

CAS RN 135926-94-2

IUPAC Name: 4,4'-dihexyl-2,2'-bithiophene
Synonyms: 4,4'-dihexyl-2,2'-bithiophene
Molecular Weight (g/mol): 334.58
Molecular Formula: C20H30S2
InChi Key: RYQPWKFBXWPBGB-UHFFFAOYSA-N
SMILES: CCCCCCC1=CSC(=C1)C1=CC(CCCCCC)=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results