accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHOO

CAS RN 135944-09-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-phenylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: CIHPCIUGLIZADU-UHFFFAOYNA-N
SMILES: OC(=O)C(CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results