accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13596-35-5

IUPAC Name: rhenium(5+) pentachloride
Synonyms: rhenium pentachloride
Molecular Weight (g/mol): 363.46
Molecular Formula: Cl5Re
InChi Key: XZQYTGKSBZGQMO-UHFFFAOYSA-I
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Re+5]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)