accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135984-01-9

IUPAC Name: N'-(phenylamino)-N-(phenylimino)thiophene-2-carboximidamide
Synonyms: N'-(phenylamino)-N-(phenylimino)thiophene-2-carboximidamide
Molecular Weight (g/mol): 306.39
Molecular Formula: C17H14N4S
InChi Key: YYNVZGCGVPELJN-UHFFFAOYSA-N
SMILES: N(N=C(N=NC1=CC=CC=C1)C1=CC=CS1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results