accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135989-69-4

IUPAC Name: 5-benzoylbenzene-1,2,4-tricarboxylic acid
Synonyms: 5-benzoylbenzene-1,2,4-tricarboxylic acid
Molecular Weight (g/mol): 314.25
Molecular Formula: C16H10O7
InChi Key: OGQPSIOWWYARAZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C(O)=O)=C(C=C1C(=O)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results